missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYRK1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599282
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DYR1B.
DYRK1B is a member of the DYRK family of protein kinases and is proposed to be involved in nuclear functions. Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. DYRK1B encodes the integrin beta 4 subunit, a receptor for the laminins. Mutations in DYRK1B are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for DYRK1B.
Specifications
| DYRK1B | |
| Polyclonal | |
| Unconjugated | |
| DYRK1B | |
| AOMS3; dual specificity tyrosine phosphorylation regulated kinase 1B; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; dual specificity tyrosine-phosphorylation-regulated kinase 1B; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; DYRK1B; DYRKB; Minibrain-related kinase; Mirk; Mirk protein kinase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13549, 9149 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y463, Q9Z188 | |
| DYRK1B | |
| A synthesized peptide derived from human DYRK1B(Accession Q9Y463), corresponding to amino acid residues M336-P386. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction