missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DYRK1A Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | DYRK1A |
|---|---|
| Dilution | Western Blot : 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence : 1-10 μg/mL, Immunoprecipitation |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067153
|
Novus Biologicals™
NBP3-30799-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066929
|
Novus Biologicals™
NBP3-30799-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DYRK1A Polyclonal antibody specifically detects DYRK1A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| DYRK1A | |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Sheep | |
| Protein Kinase | |
| Phosphate buffered saline | |
| 1859 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence : 1-10 μg/mL, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A, DYRK1, MNBEC 2.7.110EC 2.7.12 | |
| Sheep were immunized with a recombinant protein corresponding to amino acids 486 to 763 of the mouse DYRK1Aprotein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title