missing translation for 'onlineSavingsMsg'
Learn More

Dypnone, Thermo Scientific Chemicals

Product Code. 16687042 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 16687042

Brand: Thermo Scientific Acros 465850050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-45-4
Molecular Formula C16H14O
Molecular Weight (g/mol) 222.29
MDL Number MFCD00043705
InChI Key PLELHVCQAULGBH-OUKQBFOZSA-N
Synonym beta-Methylchalcone; beta-Methylstyryl phenyl ketone
IUPAC Name (2E)-1,3-diphenylbut-2-en-1-one
SMILES C\C(=C/C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Alpha Vector DYPNONE
Color Colorless to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
Solubility Information insoluble aq. soln.
Formula Weight 222.29
Physical Form Liquid
Chemical Name or Material Dypnone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.