missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dynole 34-2

Product Code. 18025202
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18025202 10 mg 10mg
18014112 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025202 Supplier Tocris Bioscience™ Supplier No. 4222/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dynamin I inhibitor

Dynamin I inhibitor (IC50 = 1.3 μM). Inhibits receptor-mediated endocytosis (RME) (IC50 = 5.0 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128165-88-7
Molecular Formula C25H36N4O

Specifications

Quantity 10 mg
Formula Weight 408.58
Purity >99%
Molecular Weight (g/mol) 408.59
Chemical Name or Material Dynamin Inhibitors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.