missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNLT1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92439-0.02ml
This item is not returnable.
View return policy
Description
DYNLT1 Polyclonal antibody specifically detects DYNLT1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DYNLT1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| CW-1, dynein light chain Tctex-type 1, dynein, light chain, Tctex-type 1, MGC111571, Protein CW-1, T-complex testis-specific protein 1 homolog, t-complex-associated-testis-expressed 1-like 1, TCTEL1tctex-1, TCTEX1, TCTEX-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human DYNLT1 (NP_006510.1). MEDYQAAEETAFVVDEVSNIVKEAIESAIGGNAYQHSKVNQWTTNVVEQTLSQLTKLGKPFKYIVTCVIMQKNGAGLHTASSCFWDSSTDGSCTVRWENKTMYCIVSAFGLSI | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6993 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction