missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNLL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16811-1-AP-20UL
This item is not returnable.
View return policy
Description
The directed movement of membrane-bounded organelles is an important process for various cellular functions including membrane transport, secretion, and axonal transport. Cytoplasmic dynein, a ubiquitous minus end-directed microtubule-based motor protein, is believed to be responsible for the retrograde transport of membranous organelles from the synapse to the cell body in fast axonal transport. The cytoplasmic dynein complex is composed of two ∽530 kDa heavy chains, and intermediate chains of ∽74 kDa and ∽53-59 kDa.Specifications
| DYNLL2 | |
| Polyclonal | |
| Unconjugated | |
| DYNLL2 | |
| 8 kDa dynein light chain b, Dlc2, DLC8b, DNCL1B, Dynein light chain LC8 type 2, DYNLL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140734, 140735, 68097 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q78P75, Q96FJ2, Q9D0M5 | |
| Dynll2 | |
| DYNLL2 Fusion Protein Ag10345 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction