missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100492
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DYNLL2.
The directed movement of membrane-bounded organelles is an important process for various cellular functions including membrane transport, secretion, and axonal transport. Cytoplasmic dynein, a ubiquitous minus end-directed microtubule-based motor protein, is believed to be responsible for the retrograde transport of membranous organelles from the synapse to the cell body in fast axonal transport. The cytoplasmic dynein complex is composed of two ~530 kDa heavy chains, and intermediate chains of ~74 kDa and ~53-59 kDa.
Specifications
| Dynein | |
| Polyclonal | |
| Unconjugated | |
| DYNLL2 | |
| 1700064A15Rik; 6720463E02Rik; 8 kDa dynein light chain; 8 kDa dynein light chain b; 8kD LC; 8kDa LC; 8kDLC; C87222; CDLC2; cytoplasmic dynein light polypeptide; cytoplasmic dynein light polypeptide 1; cytoplasmic light-chain dynein; cytosolic DH IC-1; DLC1; Dlc2; DLC8; DLC8b; DNCL1; DNCL1B; Dnclc1; dynein LC8; dynein light chain 1, cytoplasmic; Dynein light chain 2, cytoplasmic; dynein light chain LC8-type 1; Dynein light chain LC8-type 2; dynein light chain-2; dynein, cytoplasmic, light chain 1; dynein, cytoplasmic, light peptide; dynein, cytoplasmic, light polypeptide 1; dynein, light chain, LC8-type 1; dynein, light chain, LC8-type 2; DYNLL1; DYNLL2; HDLC1; LC8; LC8; PIN; LC8a; LOC540369 protein; mPIN; PIN; Protein inhibitor of neuronal nitric oxide synthase; protein inhibitor of nitric oxide synthase; radial spoke 22 homolog; RSPH22; similar to cytoplasmic dynein light chain 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140734, 140735, 56455, 58945, 68097, 8655 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P63167, P63168, P63170, Q78P75, Q96FJ2, Q9D0M5 | |
| DYNLL1, DYNLL2 | |
| A synthesized peptide derived from human DYNLL2(Accession Q96FJ2), corresponding to amino acid residues M1-K44. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction