missing translation for 'onlineSavingsMsg'
Learn More

Dynein light chain 2a cytoplasmic Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18321054 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321054 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321054 Supplier Novus Biologicals Supplier No. NBP310681100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Dynein light chain 2a cytoplasmic Polyclonal specifically detects Dynein light chain 2a cytoplasmic in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynein light chain 2a cytoplasmic
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias BITH, Bithoraxoid-like protein, DNCL2Acytoplasmic dynein light chain 2A, DNLC2ABLP, Dynein light chain 2A, cytoplasmic, dynein light chain roadblock-type 1, dynein, cytoplasmic, light polypeptide 2A, dynein, light chain, roadblock-type 1, dynein-associated protein HKM23, Dynein-associated protein Km23, roadblock domain containing 1, Roadblock domain-containing protein 1, ROBL/LC7-like 1, ROBLD1Roadblock-1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human Dynein light chain 2a cytoplasmic (NP_054902). Peptide sequence EVEETLKRLQSQKGVQGIIVVNTEGIPIKSTMDNPTTTQYASLMHSFILK
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83658
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.