missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dynein, cytoplasmic 2, light intermediate chain 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
dynein, cytoplasmic 2, light intermediate chain 1 Polyclonal specifically detects dynein, cytoplasmic 2, light intermediate chain 1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | dynein, cytoplasmic 2, light intermediate chain 1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | CGI-60, cytoplasmic dynein 2 light intermediate chain 1, D2LICDKFZp564A033, DKFZP564A033, dynein, cytoplasmic 2, light intermediate chain 1, LIC3Dynein 2 light intermediate chain |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human dynein, cytoplasmic 2, light intermediate chain 1 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?