missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC2H1 Polyclonal Antibody

DYNC2H1 Polyclonal Antibody
Brand: Invitrogen PA589005
This item is not returnable.
View return policy
Description
DYNC2H1 Polyclonal Antibody for Western Blot
Specifications
| DYNC2H1 | |
| Polyclonal | |
| Unconjugated | |
| DYNC2H1 | |
| 4432416O06Rik; AI448217; ATD3; b2b414Clo; Cytoplasmic dynein 2 heavy chain; Cytoplasmic dynein 2 heavy chain 1; cytoplasmic dynein heavy chain 2; D030010H02Rik; D330044F14Rik; DHC1b; DHC2; Dlp4; Dnch2; Dnchc2; DYH1B; Dync2h1; dynein cytoplasmic 2 heavy chain 1; dynein cytoplasmic heavy chain 2; Dynein heavy chain 11; dynein heavy chain 2; dynein heavy chain isotype 1B; dynein heavy chain, isotype 1B; dynein, cytoplasmic 2, heavy chain 1; dynein, cytoplasmic, heavy chain 2; dynein, cytoplasmic, heavy polypeptide 2; dynein-like protein 4; hDHC11; KIAA1997; mDHC11; mKIAA1997; SRPS2B; SRTD3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 65209 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.44 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9JJ79 | |
| DYNC2H1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human DYNC2H1 (NP_0010739321). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur