missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC1LI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18885-1-AP-150UL
This item is not returnable.
View return policy
Description
Cytoplasmic dynein is a microtubule-associated motorprotein.Specifications
| DYNC1LI2 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1LI2 | |
| DNCLI2, DYNC1LI2, LIC 2, LIC2, LIC53/55 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1783, 234663, 81655 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43237, Q62698, Q6PDL0 | |
| DYNC1LI2 | |
| DYNC1LI2 Fusion Protein Ag13452 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction