missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYNC1LI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525392
This item is not returnable.
View return policy
Description
This antibody is predicted to react with non-human primate based on sequence homology.
Cytoplasmic dynein is a microtubule-associated motorprotein.
Specifications
| DYNC1LI2 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1LI2 | |
| AA409702; C920003I06; cytoplasmic dynein 1 light intermediate chain 2; Dncli2; Dnclic2; Dync1li2; dynein cytoplasmic 1 light intermediate chain 2; dynein light intermediate chain 2, cytosolic; dynein, cytoplasmic 1 light intermediate chain 2; dynein, cytoplasmic 1, light intermediate chain 2; dynein, cytoplasmic, light intermediate chain 2; dynein, cytoplasmic, light intermediate polypeptide 2; fc08b04; hypothetical protein LOC550362; LIC2; LIC-2; LIC-2 dynein light intermediate chain 53/55; LIC53/55; wu:fc08b04; zgc:112480 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1783 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O43237 | |
| DYNC1LI2 | |
| KLH conjugated synthetic peptide between 152-180 amino acids from the central region of human DYNC1LI2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction