missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC1LI1 Antibody (OTI2E8), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71904PEATT594
This item is not returnable.
View return policy
Description
DYNC1LI1 Monoclonal specifically detects DYNC1LI1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| DYNC1LI1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| DYNC1LI1 | |
| Full length recombinant protein of human DYNC1LI1 (NP_057225) produced in HEK293T cells. | |
| 0.1 mL | |
| 51143.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI2E8 | |
| Flow Cytometry | |
| cytoplasmic dynein 1 light intermediate chain 1, DNCLI1, dynein, cytoplasmic 1, light intermediate chain 1, LIC1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction