missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC1I1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13808-1-AP-20UL
This item is not returnable.
View return policy
Description
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. The intermediate chains mediate the binding of dynein to dynactin via its 150 kDa component (p150-glued) DCNT1. May play a role in mediating the interaction of cytoplasmic dynein with membranous organelles and kinetochores.Specifications
| DYNC1I1 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1I1 | |
| DH IC 1, DNCI1, DNCIC1, DYNC1I1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13426, 1780, 29564 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14576, O88485, Q63100 | |
| DYNC1I1 | |
| DYNC1I1 Fusion Protein Ag4810 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction