missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYNC1I1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563981
This item is not returnable.
View return policy
Description
Immunogen sequence: LQSDSELGRR LHKLGVSKVT QVDFLPREVV SYSKETQTPL ATHQSEEDEE DEE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. The intermediate chains mediate the binding of dynein to dynactin via its 150 kDa component (p150-glued) DCNT1. May play a role in mediating the interaction of cytoplasmic dynein with membranous organelles and kinetochores.
Specifications
| DYNC1I1 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1I1 | |
| Cytoplasmic dynein 1 intermediate chain 1; cytoplasmic dynein intermediate chain 1; cytoplasmic dynein intermediate chain 1 isoform 1.6; DH IC-1; DHIC-1; DIC; Dnci 1; DNCI1; Dncic1; Dync1i1; dynein cytoplasmic 1 intermediate chain 1; dynein intermediate chain 1, cytosolic; Dynein intermediate chain 1, cytosolic (DH IC-1) (Cytoplasmic dynein intermediate chain 1); dynein, cytoplasmic 1, intermediate chain 1; dynein, cytoplasmic, intermediate chain 1; dynein, cytoplasmic, intermediate polypeptide 1; IC74 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14576 | |
| DYNC1I1 | |
| Recombinant protein corresponding to Human DYNC1I1. Recombinant protein control fragment (Product #RP-103419). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction