missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYNC1I1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529504
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, NCI-H929. Predicted reactivity: Mouse (96%), Rat (96%), Rhesus Monkey (99%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. The intermediate chains mediate the binding of dynein to dynactin via its 150 kDa component (p150-glued) DCNT1. May play a role in mediating the interaction of cytoplasmic dynein with membranous organelles and kinetochores.
Specifications
| DYNC1I1 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1I1 | |
| Cytoplasmic dynein 1 intermediate chain 1; cytoplasmic dynein intermediate chain 1; cytoplasmic dynein intermediate chain 1 isoform 1.6; DH IC-1; DHIC-1; DIC; Dnci 1; DNCI1; Dncic1; Dync1i1; dynein cytoplasmic 1 intermediate chain 1; dynein intermediate chain 1, cytosolic; Dynein intermediate chain 1, cytosolic (DH IC-1) (Cytoplasmic dynein intermediate chain 1); dynein, cytoplasmic 1, intermediate chain 1; dynein, cytoplasmic, intermediate chain 1; dynein, cytoplasmic, intermediate polypeptide 1; IC74 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.74 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O14576 | |
| DYNC1I1 | |
| Recombinant protein encompassing a sequence within the center region of human DYNC1I1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction