missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYNC1H1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568173
This item is not returnable.
View return policy
Description
Predicted to react with Human and Rat samples.
DYNC1H1 (Dynein Cytoplasmic 1 Heavy Chain 1) is a gene that encodes a protein belonging to the dynein family, which are motor proteins critical for various cellular processes. Dyneins are responsible for retrograde intracellular transport along microtubules, meaning they move cargo from the periphery of the cell towards the nucleus. The DYNC1H1 protein is a heavy chain component of cytoplasmic dynein 1, which plays a key role in the movement of organelles, vesicles, and other cellular components. Mutations in DYNC1H1 have been associated with several neurological disorders, including spinal muscular atrophy with lower extremity predominance (SMA-LED) and Charcot-Marie-Tooth disease type 2O (CMT2O). Research into DYNC1H1 continues to uncover its involvement in cellular mechanics and its implications in various genetic conditions.
Specifications
| DYNC1H1 | |
| Polyclonal | |
| Unconjugated | |
| dync1h1 | |
| 9930018I23Rik; AI894280; CMT2O; cytoplasmic dynein 1 heavy chain 1; Cytoplasmic dynein heavy chain 1; cytoplasmic dynein heavy chain polypeptide 1; Dhc1; DHC1a; DNCH1; Dnchc1; DNCL; Dnec1; DNECL; DYHC; DYNC1H1; dynein cytoplasmic 1 heavy chain 1; dynein heavy chain, cytosolic; dynein heavy chain, retrograde transport; dynein, cytoplasmic 1, heavy chain 1; dynein, cytoplasmic, heavy chain 1; dynein, cytoplasmic, heavy polypeptide 1; fk70a07; HL-3; KIAA0325; legs at odd angle; Loa; MAP 1C; MAP1C; mKIAA0325; p22; SMALED1; sprawling; Swl; wu:fk70a07 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13424, 1778, 29489 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P38650, Q14204, Q9JHU4 | |
| dync1h1 | |
| Synthesized peptide derived from human DYNC1H1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction