missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyNAzyme I & II DNA Polymerases

Product Code. 10544219 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1000U (2U/µL)
250U (2U/µL)
Unit Size:
1000 units
250 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10544219 1000U (2U/µL) 1000 units
10266269 250U (2U/µL) 250 units
10104529 1000U (2U/µL) 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10544219 Supplier Thermo Scientific™ Supplier No. F501L

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Get improved thermal stability at high temperatures with these Taq-like enzymes, Thermo Scientific™ DyNAzyme™ DNA Polymerases.

DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.

Highlights

  • Improved thermal stability
  • Improved tolerance against DMSO
  • Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
  • Detergent-free buffer also available
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage DyNAzyme II DNA Polymerase, 10x Optimized DyNAzyme BufferRecommended storage temperature -20 °C. Stable for one year from the assay date.
Description DyNAzyme™ II DNA Polymerase
Quantity 1000U (2U/µL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.