Learn More
Thermo Scientific™ DyNAzyme I & II DNA Polymerases
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1000U (2U/µL)
250U (2U/µL)
Unit Size:
1000 units
250 units
Description
DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.
Highlights
- Improved thermal stability
- Improved tolerance against DMSO
- Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
- Detergent-free buffer also available
Specifications
Specifications
| Content And Storage | DyNAzyme II DNA Polymerase, 10x Optimized DyNAzyme Buffer. Recommended storage temperature -20 °C. Stable for one year from the assay date. |
| Description | DyNAzyme™ II DNA Polymerase |
| Quantity | 250U (2U/µL) |