Learn More
Thermo Scientific™ DyNAzyme™ EXT DNA Polymerase
Product Code. 10780360 Shop All Thermo Scientific Products
Change view
Click to view available options
:
1000U (1U/uL
Product Code.
10780360 1000U (1U/uL
1 options
Product Code. 10780360 Supplier Thermo Scientific™ Supplier No. F505L
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Handle demanding reaction conditions without time-consuming optimization with this versatile and easy-to-use enzyme, Thermo Scientific™ DyNAzyme™ EXT DNA Polymerase.
Thermo Scientific™ DyNAzyme™ EXT DNA Polymerase is a versatile and easy-to-use enzyme especially suited for handling demanding reaction conditions without time-consuming optimization
It is an optimized mixture of DyNAzyme II DNA Polymerase and a proofreading PCR enzyme. DyNAzyme EXT DNA Polymerase offers exceptional performance that Taq-based mixtures cannot match.
  • Ideal for difficult templates, such as GC rich and looped sequences
  • Improved fidelity compared to standard PCR enzymes
  • Generates fragments up to 40 kb in long PCR reactions
  • High yields, robust reactions with standard templates
  • More thermostable than Taq DNA polymerase
DyNAzyme EXT DNA Polymerase is supplied with 10x Optimized DyNAzyme EXT Buffer, 10x Mg2+-free DyNAzyme EXT Buffer, 50 mM MgCl2 -solution, DMSO. Optimized DyNAzyme EXT Buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended for:
Difficult templates, long-range PCR, DHPLC, microarray, cloning
TRUSTED_SUSTAINABILITY
Specifikationer
Content And Storage 10x Optimized DyNAzyme EXT Buffer, 10x Mg2+-free DyNAzyme EXT Buffer, 50 mM MgCl2 -solution, DMSO. Optimized DyNAzyme EXT Buffer provides 1.5 mM MgCl2 in the final 1x concentration. Recommended storage temperature -20°C. Stable for one year from the assay date.
Description DyNAzyme EXT DNA Polymerase
Quantity 200U (1U/µL)
For Use With (Application) Microarray Analysis
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.