Learn More
Thermo Scientific PCR Buffer; Standard PCR; Thermo Scientific; Mg2+-free DyNAzyme Buffer and 50mM MgCl2 solution; 1x buffer contains 10mM Tris-HCl, 50mM KCl and 0.1% Triton X-100; 2 x 1.5mL of 50mM MgCl2 solution; 4 x 1.5mL of buffer
Brand: Thermo Scientific F-510L
PCR Buffer; Standard PCR; Thermo Scientific; Mg2+-free DyNAzyme Buffer and 50mM MgCl2 solution; 1x buffer contains 10mM Tris-HCl, 50mM KCl and 0.1% Triton X-100; 2 x 1.5mL of 50mM MgCl2 solution; 4 x 1.5mL of buffer
Code : A0
Additional Details : Weight : 1.51950kg