Learn More
Dynatech Precision Sampling Heavy Duty Purge and Trap Syringe from Restek
Heavy-duty glass barrel with metal front and rear flanges.
Brand: Restek 21209
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- PTFE luer lock tip
- Can fill and empty sparge tubes
- Sample-Lok valve accepts standard luer lock needles
- Suitable for purge and trap applications
Specifications
PTFE (Tip) | |
Not Applicable | |
Ea. | |
Luer Lock |
For purge and trap applications | |
Syringe | |
10 mL |