Learn More
Dynatech Precision Sampling Heavy Duty Purge and Trap Syringe from Restek
Description
- PTFE luer lock tip
- Can fill and empty sparge tubes
- Sample-Lok valve accepts standard luer lock needles
- Suitable for purge and trap applications
Specifications
Specifications
| Material | PTFE (Tip) |
| For Use With (Application) | For purge and trap applications |
| Includes | Sample lok valve |
| Product Type | Syringe |
| Quantity | Ea. |
| Volume (Metric) | 10 mL |
| Connection Type | Luer Lock |