missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dynasore
GREENER_CHOICE

Product Code. 18015142
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015142 10 mg 10mg
18095242 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015142 Supplier Tocris Bioscience™ Supplier No. 2897/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-competitive dynamin inhibitor

Dynasore is a non-competitive reversible inhibitor of dynamin 1, dynamin 2 and mitochondrial dynamin (Drp1) GTPase activity. Dynasore blocks endocytic pathways dependent on dynamin and inhibits cell spreading and migration of BSC1 cells. Dynasore attenuates motor symptoms in an animal model of Parkinson's disease and in a cellular model of Alzheimer's disease it reduces tau protein aggregation and internalization.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304448-55-3
Molecular Formula C18H14N2O4

Specifications

Quantity 10 mg
Formula Weight 322.32
Purity >99%
Molecular Weight (g/mol) 322.32
Chemical Name or Material Dynamin Inhibitors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.