missing translation for 'onlineSavingsMsg'
Learn More

DyNAmo Flash SYBR™ DyNAmo Flash SYBR Green qPCR Kit

Developed for fast real-time qPCR, the kit provides qPCR results faster than most SYBR Green kits without compromising the qPCR performance.

Brand:  Thermo Scientific™ F415S

Code : 87

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10751120

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

A DNA-binding domain attached to the polymerase in this kit improves the physical stability of the polymerase-DNA complex, thus enhancing the processivity and robustness of the amplification. The high amplification efficiency of DyNAmo Flash SYBR Green qPCR Kit gives reliable quantification and early Cq values. Due to the high signal intensity, DyNAmo Flash SYBR Green qPCR provides an excellent signal-to-noise ratio.

  • Extremely fast protocols: Combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • Included dUTP allows the use of UNG for prevention of carry-over contamination
  • Convenient 2X master mix and optimized protocol
Specifications

Specifications

Contains:
• 1 x 1 mL DyNAmo Flash SYBR Green qPCR Mix (2X)
• 1 x 50 μL 50X ROX dye, 25μM

Store at -20°C
Tube
qPCR
Fast
DyNAmo Flash SYBR Green qPCR Kit
Real Time PCR SYBR Master Mix
Gene Expression
100 reactions
1 x 1 mL
SYBR
High
Tbr DNA polymerase
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
DyNAmo Flash
100 Reactions of 20 μL, 40 Reactions of 50 μL
2X (DyNAmo Flash SYBR Green qPCR Mix), 50X (ROX Dye)
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.