Learn More
DyNAmo Flash DyNAmo Flash Probe qPCR Kit
Perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput.
Brand: Thermo Scientific™ F455S
Code : 87
Additional Details : Weight : 1.87330kg
Includes: DyNAmo Flash qPCR Mix (2X); 50X ROX dye, 25μM
Description
DyNAmo Flash Probe qPCR Kit is the perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput. Significant time savings are achieved with both conventional and fast real-time PCR instruments. Performance of the DyNAmo Flash Probe qPCR Kit is based on an efficient hot start Thermus brockianus DNA polymerase. Sensitivity and the excellent amplification efficiency across a wide range of template concentrations guarantee reproducible results.
DyNAmo Flash Probe qPCR Kit is optimized for hydrolysis probes (e.g., TaqMan probes) and it can be used with both block- and capillary-based realtime PCR instruments.
- Extremely fast protocols: combined annealing and extension step of only 15 sec.
- Specific and sensitive detection of a wide range of template concentrations
- Included dUTP allows the use of UNG for prevention of carry-over contamination
- Convenient 2X master mix and optimized protocols
Specifications
Contains: • 1 x 1 mL DyNAmo Flash qPCR Mix (2X) • 1 x 50 μL 50X ROX dye, 25μM Store at -20°C |
|
Tube | |
qPCR | |
Fast | |
DyNAmo Flash Probe qPCR Kit | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
100 Reactions | |
1 x 1 mL |
Primer-probe | |
High | |
Tbr DNA polymerase | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Stratagene Mx4000, StepOne™ System, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, BioRad MyiQ, BioRad iQ5, BioRad iCycler iQ, Cepheid SmartCycler, Stratagene Mx3005P | |
DyNAmo Flash | |
100 Reactions of 20 μL, 40 Reactions of 50 μL | |
2X (DyNAmo Flash qPCR Mix), 50X (ROX Dye) | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.