missing translation for 'onlineSavingsMsg'
Learn More

DyNAmo ColorFlash DyNamo ColorFlash Probe qPCR Kit

Offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting.

Brand:  DyNAmo ColorFlash F456L

 View more versions of this product

Product Code. 10401358

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: DyNAmo ColorFlash qPCR Mix (2X); 50X ROX dye, 25μM; 40X sample buffer solution with yellow dye
This item is not returnable. View return policy

Description

Description

DyNAmo ColorFlash Probe qPCR Kit offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting. The 2X master mix contains a blue dye, and a separate sample buffer contains a yellow dye. The qPCR reaction mix containing both components is green. Using this patent-pending multicolor system, pipetting of both the master mix and the sample can be easily monitored. This significantly decreases the risk for pipetting errors during reaction set up, especially when using white reaction vessels. The dyes do not affect the specificity or sensitivity of qPCR assays.

  • Minimized risk of pipetting errors during reaction set up
  • Especially beneficial when using white reaction vessels
  • Extremely fast protocols due to combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • dUTP included in the 2X master mix allows the use of UNG for prevention of carry-over contamination
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 5 x 1 mL DyNAmo ColorFlash qPCR Mix (2X)
• 1 x 250 μL 50X ROX dye, 25μM
• 1 x 1000 μL 40X sample buffer solution with yellow dye

Store at -20°C
Tube
qPCR
Fast
DyNAmo ColorFlash Probe qPCR Kit
Real Time PCR SYBR Master Mix
Gene Expression
500 Reactions
5 x 1 mL
Primer-probe
High
Tbr DNA polymerase
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Stratagene Mx4000, StepOne™ System, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, BioRad MyiQ, BioRad iQ5, BioRad iCycler iQ, Cepheid SmartCycler, Stratagene Mx3005P
DyNAmo Flash
500 Reactions of 20 μL, 200 reactions of 50 μL
2X (DyNAmo ColorFlash qPCR Mix), 50X (ROX Dye), 40X (Sample Buffer Solution with Yellow Dye)
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.