missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamitin (1-86 AA) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10032-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is against the N terminal 86aa of full length p50 dynamitin.
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.Specifications
| Dynamitin (1-86 AA) | |
| Polyclonal | |
| Unconjugated | |
| DCTN2 | |
| DCTN 50, DCTN2, DCTN50, dynactin 2, dynactin 2 (p50), Dynactin subunit 2, DYNAMITIN, p50 dynamitin, RBP50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10540, 69654 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13561, Q99KJ8 | |
| Dctn2 | |
| Dynamitin (1-86 AA) Fusion Protein Ag0029 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction