missing translation for 'onlineSavingsMsg'
Learn More

Dynamin 3 Rabbit anti-Human, Mouse, Clone: 5X7T4, Novus Biologicals™

Product Code. 18332492 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332492 20 μg 20µL
18374084 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18332492 Supplier Novus Biologicals Supplier No. NBP31572220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Dynamin 3 Monoclonal antibody specifically detects Dynamin 3 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynamin 3
Applications Western Blot
Classification Monoclonal
Clone 5X7T4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Dyna III, dynamin 3, dynamin family member, dynamin-3, EC 3.6.5.5, MGC70433, T-dynamin, testicular
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 700-800 of human Dynamin 3 (Q9UQ16).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 26052
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.