missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynamin 3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142348
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
The dynamins are a family of 100 kDa GTPases transcribed from at least three separate genes. At least four mRNA splice variants for each dynamin have been described. Dynamins contain several conserved regions including the conserved, amino-terminal GTPase domain, a centrally located membrane-binding plekstrin homology domain (PHD), and a coiled-coil region located in front of a proline-rich domain (PRD). The PRD is thought to mediate interactions between dynamin and numerous other cellular proteins. Dynamin I is expressed exclusively in neurons, dynamin II is ubiquitously expressed, and dynamin III is thought to be restricted to expression in the brain, testis, heart, and lung. The dynamins participate in the cellular process of clathrin-mediated and fluid-phase endocytosis.
Specifications
| Dynamin 3 | |
| Polyclonal | |
| Unconjugated | |
| Dnm3 | |
| 9630020E24Rik; AW061159; B230343F03Rik; C530045C17; DNM3; Dyn3; Dyna III; dynamin 3; dynamin family member; Dynamin III; dynamin, testicular; dynamin-2; Dynamin-3; KIAA0820; mKIAA0820; T-dynamin; testicular dynamin | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 26052 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UQ16 | |
| Dnm3 | |
| Peptide with sequence C-DKSVGNNKAEND. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction