missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin 3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92627-0.1ml
This item is not returnable.
View return policy
Description
Dynamin 3 Polyclonal antibody specifically detects Dynamin 3 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Dynamin 3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Dyna III, dynamin 3, dynamin family member, dynamin-3, EC 3.6.5.5, MGC70433, T-dynamin, testicular | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 410-555 of human Dynamin 3 (NP_001265181.1). EAIVKKQIVKLKGPSLKSVDLVIQELINTVKKCTKKLANFPRLCEETERIVANHIREREGKTKDQVLLLIDIQVSYINTNHEDFIGFANAQQRSSQVHKKTTVGNQVIRKGWLTISNIGIMKGGSKGYWFVLTAESLSWYKDDENI | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 26052 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction