missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynamin 2 Recombinant Rabbit Monoclonal Antibody (23GB5170), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Dynamin 2 |
|---|---|
| Clone | 23GB5170 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089856
![]() |
Invitrogen™
MA553858 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087920
|
Invitrogen™
MA553859 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dynamin 2 Recombinant Monoclonal antibody specifically detects Dynamin 2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Dynamin 2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P39052, P39054, P50570 | |
| 13430, 1785, 25751 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5170 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; Dnm2; DYIIAAB; Dyn2; dynamin 2; dynamin II; Dynamin UDNM; Dynamin-2; DYNII; LCCS5; Udnm | |
| A synthesized peptide derived from human Dynamin 2 (750-800AA). | |
| Primary | |
| -20°C | |
| DNM2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title