missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynamin 1 Recombinant Rabbit Monoclonal Antibody (23GB5165), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Dynamin 1 |
|---|---|
| Clone | 23GB5165 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089422
|
Invitrogen™
MA552806 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089107
|
Invitrogen™
MA552807 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dynamin 1 Recombinant Monoclonal antibody specifically detects Dynamin 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| Dynamin 1 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P39053, Q05193 | |
| 13429, 1759 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5165 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI838169; B-dynamin; D100; Dnm; Dnm1; Dyn1; dynamin 1; Dynamin BREDNM19; dynamin i; dynamin, brain; Dynamin1; Dynamin-1; EC 3.6.5.5; EIEE31; Ftfl; Kiaa4093; mKIAA4093; Neuronal dynamin; OTTHUMP00000215500 | |
| A synthesized peptide derived from human Dynamin 1 (800-864AA). | |
| Primary | |
| -20°C | |
| DNM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title