missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18205-1-AP-150UL
This item is not returnable.
View return policy
Description
Microtubule-associated force-producing protein involved in producing microtubule bundles and able to bind and hydrolyze GTP. Most probably involved in vesicular trafficking processes. Involved in receptor-mediated endocytosis.Specifications
| Dynamin 1 | |
| Polyclonal | |
| Unconjugated | |
| DNM1 | |
| DNM, DNM1, dynamin 1, Dynamin1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13429, 140694, 1759 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21575, P39053, Q05193 | |
| DNM1 | |
| Dynamin 1 Fusion Protein Ag12827 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction