missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynamin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5122022

 View more versions of this product

Product Code. 17703424

  • £155.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Dynamin 1 Polyclonal Antibody for Western Blot, IHC (P)

Dynamin is a GTPase required for endocytosis of clathrin coated vesicles from the membrane. Dynamin's involvement in endocytosis was first discovered during the characterization of the temperature sensitive parylytic shibire mutation from Drosophila melanogaster. Three closely related isoforms have been identified in mammals. Dynamin1 is expressed exclusively in neurons, dynamin2 is ubiquitously expressed and dynamin3 is expressed primarily in the testes. Each of these isoforms of dynamin also has several splice variants. This suggests that each isoform/splice variant may have separate intracellular targets or function in the budding of distinct vesicle types. Common domain features of the dynamin proteins include an amino acid terminal GTPase domain, a central pleckstrin homology domain and a highly basic proline rich carboxyl terminus with several Src homology 3 (SH3) domains. These SH3 domains target the dynamin to the clathrin coated pit. Dynamin is known to interact with at least two proteins, AP2 adaptor protein complex and amphiphysin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dynamin 1
Polyclonal
Unconjugated
DNM1
AI838169; B-dynamin; D100; Dnm; Dnm1; Dyn1; dynamin 1; Dynamin BREDNM19; dynamin i; dynamin, brain; Dynamin1; Dynamin-1; EC 3.6.5.5; EIEE31; Ftfl; Kiaa4093; mKIAA4093; Neuronal dynamin; OTTHUMP00000215500
Rabbit
Antigen affinity chromatography, Protein A
RUO
13429, 140694, 1759
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P21575, P39053, Q05193
DNM1
Recombinant protein Dynamin 1. The antigen corresponds to amino acid range 572-745 of the target protein.
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.