Learn More
ABEBA™ Dynamic Flow Anthracite Occupational Slip-On O1 ESD
Product Code. 17141405
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17141405
Brand: ABEBA™ 37375/41
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Anthracite dynamic flow occupational slip-on shoes with replaceable active MediFit insole, slip resistant TPU outsole
  • Breathable, lightweight, slip-on shoes
  • Comply to EN ISO 20347: 2012, O1 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Anthracite knitted fabric breathable
  • Double elastic band fitting system
  • Removable active Medifit insole
Specifications
Color Anthracite
Material Knitted Fabric
Size 41
Certifications/Compliance (Europe) Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340
Gender Unisex
Ankle Style Slip-on
Waterproof Water Repellent
Product Type O1 SRC ESD