Learn More
ABEBA™ Dynamic Flow Anthracite Occupational Slip-On O1
Occupational slip-on shoes, breathable anthracite knitted fabric, replaceable active MediFit insole, slip resistant TPU outsole
Brand: ABEBA™ 7375/35
Additional Details : Weight : 0.46000kg
Description
- Breathable, lightweight, comfortable
- Comply to EN ISO 20347: 2012, O1 SRC
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Anthracite knitted fabric breathable
- Double elastic band fitting system
- Removable active Medifit insole
Specifications
Anthracite | |
35 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
Comply to EN ISO 20347: 2012 O1 SRC | |
Slip-on | |
O1 SRC ESD |