Learn More
Abeba™ Dynamic 7345 Shoes
Product Code. 15662768
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15662768
Brand: Abeba™ 7345/44
Only null left
Add to basket
Add to basket
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber with breathable textile inlays
  • White/Black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White, Black
Material Microfiber
Size 44
Sole Features Slip-resistant
Sole Material Polyurethane
Style 7345