Learn More
Abeba™ Dynamic 7312 Shoes
Product Code. 15692648
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15692648 39 1 pair
15652648 35 1 pair
15662648 36 1 pair
15672648 37 1 pair
15682648 38 1 pair
15602658 40 1 pair
15612658 41 1 pair
15622658 42 1 pair
15632658 43 1 pair
15642658 44 1 pair
15652658 45 1 pair
15662658 46 1 pair
15672658 47 1 pair
15682658 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15692648 Supplier Abeba™ Supplier No. 7312/39
  • ONLINE EXCLUSIVE PRICE
    £57.85
    £52.92 / 1 pairs
    Save £4.93
    8% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/blue
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White
Material Leather
Size 39
Sole Features Slip-resistant
Sole Material Polyurethane
Style 7312