missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ DynaLight™ Substrate with RapidGlow™ Enhancer
Click to view available options
Quantity:
1 L
Unit Size:
Each
Description
Achieve faster ELISA results with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized to achieve faster results in solution-based assays. The DynaLight™ Substrate with RapidGlow™ Enhancer formulation includes 1,2-dioxetane chemiluminescent substrate and a polymeric enhancer that enables ultra-sensitive immunoassay detection by alkaline phosphatase label.
Features of DynaLight™ Substrate with RapidGlow™ Enhancer:
Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
Sensitive attomole detection of purified alkaline phosphatase enzyme
Optimized formulation for both microplate and bead-based assays
Substrate stability of over 1 year at 4°C
Compatibility with automated platforms with trigger solution/positions already in place
The DynaLight™ Substrate with RapidGlow™ Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline phosphatase. This reaction achieves maximum light emission within 2 minutes and maintains a sustained signal for hours. When combined with DynaLight™ Trigger Solution, DynaLight™ Substrate with RapidGlow™ Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight™ Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Features of DynaLight™ Substrate with RapidGlow™ Enhancer:
The DynaLight™ Substrate with RapidGlow™ Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline phosphatase. This reaction achieves maximum light emission within 2 minutes and maintains a sustained signal for hours. When combined with DynaLight™ Trigger Solution, DynaLight™ Substrate with RapidGlow™ Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight™ Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Specifications
| Description | DynaLight™ Substrate with RapidGlow™ Enhancer, 1L, Liquid, Chemiluminescent Detection Method, Cell Cultures, Serum, Whole Blood Samples, 1pg/ml Sensitivity, 2 min. Reaction Time, For Protein Assays and Analysis |
| Product Type | ELISA Substrate with Enhancer |
| Quantity | 1 L |
| Detection Method | Chemiluminescence |
| Assay Sensitivity | 1 pg/mL |
| Form | Liquid |
| For Use With (Application) | Protein biology, cell analysis, enzyme reporter assays, ELISA, protein assays and analysis, alkaline phosphatase assays, build your own immunoassay |
| Sample Type | Cell Cultures, Whole Blood, Serum |
| Test Time | 2 min. |
| Packaging Quantity | 1 Each |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction