missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaGreen™ CaptureSelect™ Anti-IgG-Fc (Multi-Species) Magnetic Beads
GREENER_CHOICE

Product Code. 17816667
Click to view available options
Quantity:
25 mL, 1000 Reactions
3 mL, 120 Reactions
Unit Size:
25mL
3mL
This item is not returnable. View return policy

Product Code. 17816667

Brand: Invitrogen™ 80109G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DynaGreen CaptureSelect Anti-IgG-Fc (Multi-Species) Magnetic Beads enable high-performance direct and indirect immunoprecipitation (IP) while also being an environmentally sustainable choice.

DynaGreen CaptureSelect Anti-IgG-Fc (Multi-Species) Magnetic Beads enable high-performance direct and indirect immunoprecipitation while also being an environmentally sustainable choice. DynaGreen CaptureSelect Anti-IgG-Fc (ms) Magnetic Beads are microplastic-free submicron magnetic beads. The beads provide reproducible isolation of target proteins in a simple and efficient workflow that can be performed manually or automated on KingFisher instruments.

Features include:

  • Generate reproducible high-yield, high-quality immunoprecipitation
  • Reduce environmental impact of research
  • Enhanced purity and low non-specific binding
  • Effortless scale-up and automation
  • Non-porous beads for low antibody consumption

Simple, gentle, and efficient manual separation

DynaGreen CaptureSelect Anti-IgG-Fc (ms) Magnetic Beads are microplastic-free superparamagnetic beads with CaptureSelect Anti-IgG-Fc ligand covalently coupled to the surface. The submicron bead size (∼250 nm) provides a low sedimentation rate and large available target capture surface area. This results in efficient, high-yield isolation of target protein by direct or indirect immunoprecipitation in less than 80 minutes in a simple bind-incubate-elute protocol, with no pre-clearing required.

The CaptureSelect Anti-IgG-Fc ligand is recombinant camelid-derived single-domain antibody fragment that specifically binds to the Fc part of IgG from multiple species, and avoids cross-binding to other isotypes, such as IgA and IgM.

The magnetic separation technology used by DynaGreen CaptureSelect Anti-IgG-Fc (ms) Magnetic Beads is also rapid and gentle , thereby causing minimal physical stress to your target proteins. Furthermore, the non-porous nature of the beads means there is no risk of antibodies getting trapped and becoming inaccessible, thereby minimizing the amount of antibody required to produce accurate and reproducible results.

The yield and purity of the isolated target protein is highly compatible for use in western blot or mass spectrometry.

The product manual below includes a list of required reagents and materials and details of compatible buffer recipes.

Rapid and reproducible automated workflow

As the scale of experiments increases to medium and high throughput, DynaGreen CaptureSelect Anti-IgG-Fc (ms) Magnetic Beads workflows can be seamlessly scaled using the KingFisher automation platforms. The automated protocol can be run in just 40 minutes with up to 96 samples per run, which significantly reduces hands-on-time while maintaining the same reproducible high target yield and low non-specific binding vs the manual protocol.

A certified choice for environmentally sustainable research

Microplastic-free DynaGreen CaptureSelect Anti-IgG-Fc (ms) Magnetic Beads were designed for both high performance in immunoprecipitation workflows and with the environment in mind. They have been shown to provide superior purity and high yield of target protein compared to other magnetic bead alternatives in both direct and indirect immunoprecipitation workflows. At the same time the product boasts an extensive list of third-party ACT-label-certified sustainable product design elements by incorporating principles of green chemistry and green engineering. Choosing DynaGreen CaptureSelect Anti-IgG-Fc (ms) Magnetic Beads thereby helps reduce the environmental impact of research without compromising on results.

TRUSTED_SUSTAINABILITY

Specifications

Green Features Less hazardous, less waste, sustainable packaging
Concentration 20 mg/mL
Material Iron Oxide
Color Golden Brown, Black
For Use With (Application) Immunoprecipitation
Product Type Antibody Coated Bead
Content And Storage DynaGreen CaptureSelect Anti-IgG-Fc (Multi-Species) Magnetic Beads are supplied in phosphate buffer, pH 7.4, with a biodegradable surfactant and a preservative. Store at 2–8°C.
Diameter (Metric) 250 nm
For Use With (Equipment) DynaMag Magnets, or KingFisher Instruments
Shipping Condition Ambient
Quantity 25 mL, 1000 Reactions
Type Magnetic Affinity Bead
Ligand Type CaptureSelect Anti-IgG-Fc
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.