missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dynactin Subunit 2/DCTN2/DCTN-50 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29034
This item is not returnable.
View return policy
Description
Dynactin Subunit 2/DCTN2/DCTN-50 Polyclonal antibody specifically detects Dynactin Subunit 2/DCTN2/DCTN-50 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| Dynactin Subunit 2/DCTN2/DCTN-50 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DCTN-50, DCTN5050 kD dynein-associated polypeptide, dynactin 2 (p50), dynactin complex 50 kD subunit, Dynactin complex 50 kDa subunit, dynactin subunit 2,50 kDa dynein-associated polypeptide, DYNAMITIN, p50 dynamitin, RBP50 | |
| between 351 and 401 | |
| 20 μg | |
| Cell Cycle and Replication | |
| 10540 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction