missing translation for 'onlineSavingsMsg'
Learn More

Dynactin Subunit 2/DCTN2/DCTN-50 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30736139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736139 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736139 Supplier Novus Biologicals Supplier No. AF6850D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Dynactin Subunit 2/DCTN2/DCTN-50 Polyclonal antibody specifically detects Dynactin Subunit 2/DCTN2/DCTN-50 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynactin Subunit 2/DCTN2/DCTN-50
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias DCTN-50, DCTN5050 kD dynein-associated polypeptide, dynactin 2 (p50), dynactin complex 50 kD subunit, Dynactin complex 50 kDa subunit, dynactin subunit 2,50 kDa dynein-associated polypeptide, DYNAMITIN, p50 dynamitin, RBP50
Host Species Sheep
Immunogen E. coli-derived recombinant human Dynactin Subunit 2/DCTN2, Asp277-Lys401, Accession # Q13561
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10540
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.