missing translation for 'onlineSavingsMsg'
Learn More

Dynactin Subunit 2/DCTN2/DCTN-50 Antibody (696206) [DyLight 680], Novus Biologicals™

Product Code. 30644679 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30644679 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30644679 Supplier Novus Biologicals Supplier No. FAB6850Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dynactin Subunit 2/DCTN2/DCTN-50 Monoclonal antibody specifically detects Dynactin Subunit 2/DCTN2/DCTN-50 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynactin Subunit 2/DCTN2/DCTN-50
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 696206
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias DCTN-50, DCTN5050 kD dynein-associated polypeptide, dynactin 2 (p50), dynactin complex 50 kD subunit, Dynactin complex 50 kDa subunit, dynactin subunit 2,50 kDa dynein-associated polypeptide, DYNAMITIN, p50 dynamitin, RBP50
Host Species Mouse
Immunogen E. coli-derived recombinant human Dynactin, Subunit 2/DCTN2, Asp277-Lys401, Accession # Q13561
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10540
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.