missing translation for 'onlineSavingsMsg'
Learn More

Dynactin Subunit 1/DCTN1 Antibody (705007) [DyLight 755], Novus Biologicals™

Product Code. 30643756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30643756 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30643756 Supplier Novus Biologicals Supplier No. FAB6657Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dynactin Subunit 1/DCTN1 Monoclonal antibody specifically detects Dynactin Subunit 1/DCTN1 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynactin Subunit 1/DCTN1
Applications Western Blot
Classification Monoclonal
Clone 705007
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), p135, p150 Glued (Drosophila) homolog, p150-glued
Host Species Mouse
Immunogen E. coli-derived recombinant human Dynactin Subunit 1/DCTN1, Ala1145-Ser1278, Accession # Q14203
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1639
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.