missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dynactin Subunit 1/DCTN1 Monoclonal antibody specifically detects Dynactin Subunit 1/DCTN1 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | Dynactin Subunit 1/DCTN1 |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1E12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | Q14203 |
| Gene Alias | dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), p135, p150 Glued (Drosophila) homolog, p150-glued |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?