missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynactin 6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92536-0.02ml
This item is not returnable.
View return policy
Description
Dynactin 6 Polyclonal antibody specifically detects Dynactin 6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Dynactin 6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dynactin 6, dynactin subunit 6, Dynactin subunit p27, P27, Protein WS-3, WS-3, WS3novel RGD-containing protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human DCTN6 (NP_006562.1). MAEKTQKSVKIAPGAVVCVESEIRGDVTIGPRTVIHPKARIIAEAGPIVIGEGNLIEEQALIINAYPDNITPDTEDPEPKPMIIGTNNVFEVGCYSQAMKMGDNNVIESKAYVGRNVILTSGCIIGACCNLNTFEVIPENTVIYGADCLRRVQTERPQPQTLQLDFLMKILPNYHHLKKTMKGSSTPVKN | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Immunology, Signal Transduction | |
| 10671 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction