missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynactin 4 Polyclonal Antibody
Dynactin 4 Polyclonal Antibody
Brand: Invitrogen PA550392
This item is not returnable.
View return policy
Description
Dynactin 4 Polyclonal Antibody for IHC (P)
Specifications
| Dynactin 4 | |
| Polyclonal | |
| Unconjugated | |
| DCTN4 | |
| 1110001K06Rik; 4930547K17Rik; C130039E17Rik; DCTN4; Dyn4; dynactin 4; dynactin 4 (p62); dynactin 4 (p62) isoform 1; dynactin 4 (p62) isoform 2; dynactin p62 subunit; dynactin subunit 4; Dynactin subunit p62; hypothetical protein LOC550479; P62; si:ch211-174h24.2; zgc:112431 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51164 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UJW0 | |
| DCTN4 | |
| Fusion protein corresponding to residues near the C terminal of human dynactin 4 (p62). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction