missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynactin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560864
This item is not returnable.
View return policy
Description
Immunogen sequence: FILSQVALLE QVNALVPMLD SAHIKAVPEH AARLQRLAQI HIQQQDQCVE ITEESKALLE EYNKTTMLLS KQFVQWDELL CQLEAATQVK PA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.
This gene encodes the smallest subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, cytokinesis, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like most other dynactin subunits, exists only as a part of the dynactin complex. It is primarily an alpha-helical protein with very little coiled coil, and binds directly to the largest subunit (p150) of dynactin. Alternative splicing of this gene generates 2 transcript variants.
Specifications
| Dynactin 3 | |
| Polyclonal | |
| Unconjugated | |
| DCTN3 | |
| DCTN22; DCTN-22; DCTN3; dynactin 3; dynactin 3 (p22); dynactin complex subunit 22 kDa subunit; dynactin light chain; dynactin light chain (P24); dynactin light chain p24; Dynactin subunit 3; MGC111190; p22; p24; p24 dynactin light chain | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75935 | |
| DCTN3 | |
| Recombinant protein corresponding to Human Dynactin 3. Recombinant protein control fragment (Product #RP-92986). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction