Learn More
Invitrogen™ Dynactin 2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547881
Description
This antibody detects human Dynactin Subunit 2/DCTN2 in direct ELISAs and human and mouse Dynactin Subunit 2/DCTN2 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human DCTN1 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.
Specifications
| Dynactin 2 | |
| Polyclonal | |
| Unconjugated | |
| DCTN2 | |
| 2310042E05Rik; 50 kDa dynein-associated polypeptide; C130077D06Rik; C87049; DCTN2; dctn2 protein; DCTN50; DCTN-50; dynactin 2; dynactin 2 (p50); dynactin complex 50 kDa subunit; Dynactin subunit 2; dynamitin; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; HEL-S-77; hypothetical protein LOC527201; p50; p50 dynamitin; p50/dynamitin; RBP50; zgc:63867 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 10540, 69654 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q13561, Q99KJ8 | |
| DCTN2 | |
| E. coli-derived recombinant human Dynactin Subunit 2/DCTN2 Asp277-Lys401. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.